Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RediLoad™ Loading Buffer

Numéro de catalogue. 750026
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
75-002-6 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 75-002-6 Fournisseur Invitrogen™ Code fournisseur 750026
  • $261.00
    $242.65 / Each of 1

    Save $18.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Afficher toutes les promotions
Ajouter au panier
Edge
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Designed for easy loading and tracking of PCR products run in agarose gels

RediLoad™ is added directly to the PCR reaction mixture prior to amplification. RediLoad features a reagent that increases density for easy loading, and a red tracking dye that migrates at approximately 25bp and allows easy monitoring of electrophoresis. RediLoad can be used as a replacement for conventional agarose loading buffer and is supplied in a 10X concentration. Compatible with agarose gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Spécifications

Content And Storage Store in refrigerator (2–8°C).
Buffer Sample Loading Buffers
Concentration 10X
Gel Type Agarose
Gel Compatibility E-Gel™, Agarose Gels
Quantity 10 mL
Shipping Condition Room Temperature
Product Line Ready-Load
Product Type Loading Buffer
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.