Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ Double-Stranded cDNA Synthesis Kit

Catalog No. p-4921680
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Reactions
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-917-010 10 Reactions
11-917-020 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-917-010 Supplier Invitrogen™ Supplier No. 11917010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50μg total RNA or 0.2-5μg mRNA into double-stranded cDNA.

Requires

Primer

Maximize the yield of full-length cDNA, as well as that of overall cDNA synthesis

Contains all of the reagents (except an oligo (dT)-containing primer) necessary to make high-quality, double-stranded cDNA from total RNA or poly A+-selected RNA (mRNA).

Simple, reliable, and trouble-free
The SuperScript Double-Stranded cDNA Synthesis Kit offers a simple, reliable, and trouble-free method by which RNA templates can be converted to cDNA for use in cloning and library construction experiments. The reactions described in the SuperScript Double-Stranded cDNA Synthesis Kit protocol are designed to convert 25–50μg total RNA, or 0.2–5μg mRNA, into first- and second-strand cDNA (supply your own primer). Enjoy the benefits of a tested protocol and reagents of consistent quality. To get the best possible cDNA product using this kit, use a high-quality RNA substrate.

  • Reliability: each buffer, reagent, and enzyme in the kit is of the same high quality you expect from Life Technologies
  • Performance: SuperScript II is an enzyme you can trust for high-yield, high-quality, general-purpose, first-strand synthesis
  • Optimal temperature: 42°C

DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis, PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: The kit includes reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50 μg total RNA or 0.2-5 μg mRNA into double-stranded cDNA.

Storage: Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Format Kit
Labeling Method Direct Labeling
Optimal Reaction Temperature 42°C
Product Line SuperScript
Product Type cDNA Synthesis Kit
Quantity 10 Reactions
Reverse Transcriptase SuperScript™ II
Shipping Condition Wet Ice
For Use With (Application) Microarray Analysis
Final Product Type cDNA (Double-Stranded)
Sample Type RNA
Show More Show Less
I'd like to generate double-stranded DNA (dsDNA) from mRNA. Do you have a kit for this?

To generate double-stranded cDNA, we recommend the Maxima H Minus Double-Stranded cDNA Synthesis Kit (Cat. No. K2561) or Second Strand cDNA Synthesis Kit (Cat. No. A48570).

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.