Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Safe Imager™ Viewing Glasses

Catalog No. S37103
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37103 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37103 Supplier Invitrogen™ Supplier No. S37103
  • $99.50
    $95.65 / Each of 1

    Save $3.85

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

I pair of Safe Imager™ viewing glasses

For use with the Safe Imager™ blue-light transilluminator

The Safe Imager viewing glasses allow visualization of gel bands without use of the Safe Imager amber filter to facilitate band excision.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage I pair of Safe Imager™ viewing glasses
For Use With (Equipment) Safe Imager™
Product Line SYBR Safe
Type Viewing Glass
Quantity 1 Each
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.