Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Topoisomerase I

Catalog No. 38042024
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-042-024 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-042-024 Supplier Invitrogen™ Supplier No. 38042024
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Catalyzes removal of superhelical turns from covalently closed DNA by a transient breakage and rejoining of phosphodiester bonds

  • Topoisomerase I is active in the presence of EDTA
  • Source: Purified from calf thymus
  • Performance and Quality Testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease, and phosphatase assays; conversion of super-coiled DNA to relaxed DNA
  • Unit Definition: One unit catalyzes the conversion of 0.5μg of superhelical ΦX174 RF DNA to a relaxed state in 30 min. at 37°C
  • Unit Reaction Conditions: 50mM Tris-HCl (pH 7.5), 50mM KCl, 10mM MgCl2 , 0.1mM EDTA, 0.5mM DTT, 30 μg/mL BSA, 0.5μg ΦX174 RF DNA, and enzyme in 50μL for 30 min. at 37°C

Relaxing positively and negatively supercoiled DNA, Producing DNA topoisomers, Cloning, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Wet Ice
Enzyme Topoisomerase
Quantity 500 U
Product Type Topoisomerase I
What are the applications and recommended protocols for Topoisomerase I?

Topoisomerase I has two general applications: to relax supercoiled DNA, and to generate samples with defined superhelical density. Generating samples with defined superhelical density is described in Analytical Biochemistry, v.122, p. 253, by Singleton and Wells (1982).

For relaxing supercoiled DNA, use the following protocol:

1. Add 0.5 µg supercoiled PhiX 174 DNA, 50 mM TrisHCl (pH 7.5), 50 mM KCl, 10 mM MgCl2, 0.5 mM DTT, 0.1 mM EDTA, 30 µg/mL BSA, and 1 unit Topoisomerase I to a final volume of 50 µL.
2. Incubate 30 mins at 37 degrees C.
3. Analyze the treated samples on a 1% agarose gel that does not contain ethidium bromide. Stain with ethidium bromide after electrophoresis is complete.

Do you offer a pre-made buffer for use with Topoisomerase I (Cat. No. 38042024)?

No, we do not offer a pre-made buffer for use with this product.

When editing a cycle on the Procise System, I can readily delete a step, but cannot seem to insert a function into the cycle. What am I doing wrong?

When you edit a flask cycle, be sure to highlight a flask function in order to insert it. Similarly, when editing a cartridge cycle, highlight a cartridge function in order to insert it.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.