Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrackIt™ Cyan/Yellow Loading Buffer
GREENER_CHOICE

Catalog No. 10482035
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-482-035 3 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-482-035 Supplier Invitrogen™ Supplier No. 10482035
  • $74.00
    $71.65 / Each of 1

    Save $2.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

6X solutions used for easy loading and tracking of DNA samples in agarose gels.

Loading buffers contain two tracking dyes, one that runs behind sample and one that runs ahead of sample. Tracking dyes serve as visual markers for following migration during electrophoresis and indicate when maximum resolution of DNA fragments has been achieved. Additionally, visualization of DNA bands will not be obscured by tracking dyes because they run outside limits of most DNA samples.

 

  • TrackIt Cyan/Orange Loading Buffer is recommended for DNA fragments between 50 bp and 10 Kb
  • TrackIt Cyan/Yellow Loading Buffer is recommended for DNA fragments between 10 bp and 1 Kb

 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 3 x 0.5 mL 6X TrackIt Cyan/Yellow Loading Buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

Buffer Sample Loading Buffers
Concentration 6X
Gel Type Agarose
Label or Dye Tartrazine, Xylene Cyanol, Xylene Cyanol FF
Sample Loading Volume 0.5 mL
Gel Compatibility Agarose Gels
Quantity 3 x 0.5 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line TrackIt
Product Type Loading Buffer
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.