Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Glycogen

Catalog No. 10814010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-814-010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-814-010 Supplier Invitrogen™ Supplier No. 10814010
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

1 vial containing 100μL of 20μg/μL ultrapure glycogen

Used as carrier for nucleic acid precipitation in ethanol

  • Concentration: 20μg/μL in RNase-free water
  • Performance and quality testing: No DNase or RNase activity detected

ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, RNAi, Epigenetics & Noncoding RNA Research

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Grade Molecular Biology
Content And Storage 1 vial containing 100 μl of 20 μg/μl ultrapure glycogen.

Store at -5°C to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 100 μL
Product Type Glycogen
Product Line UltraPure
Form Liquid
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.