Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Sodium Dodecyl Sulfate (SDS)

Catalog No. 15525017
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-525-017 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-525-017 Supplier Invitrogen™ Supplier No. 15525017
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Detergent that functions as protein denaturant

  • Used in determination of protein molecular weight by polyacrylamide gel electrophoresis
  • Used to disrupt cell walls and dissociate nucleic acid-protein complexes in DNA and RNA extraction protocols

DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Form Powder
For Use With (Application) Nucleic Acid Extraction
Quantity 500 g
Reagent Type SDS
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line UltraPure
Product Type Sodium Dodecyl Sulfate
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.