Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Wash Buffer (25X)

Catalog No. p-4926822
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WB01 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WB01 Supplier Invitrogen™ Supplier No. WB01
Only null left
Add to Cart
Edge
Add to Cart

Suitable for use with all ELISA assays

  • Clear, colorless concentratred liquid buffer
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Content And Storage Buffer. Store at 2°C to 8°C.
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.