Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodide Iodate concentrate, For 1L standard solution, 1/128 M I2 (1/64N), Solstice

Catalog No. 60046686 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-686 6 x 1 Ea.
60-026-64 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-686 Supplier Solstice Supplier No. 380646X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 1/128 M I2 (1/64N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0 , 1310-58-3 , 7758-05-6 , 7732-18-5
Molecular Formula IK
MDL Number MFCD00011355
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
SMILES [K+].[I-]

Specifications

CAS Max % 1.2%, ≥95.0000%
CAS Min % ≥95.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
UN Number NONH for all modes of transport
Beilstein 3587194
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
Packaging Ampoule
Density 1.160 g/cm3
Chemical Name or Material Iodide Iodate concentrate, For 1L standard solution, 1/128 M I2 (1/64N)
Show More Show Less
Hazard Statement P305 + P351 + P338
ShelfLife 1080 days from date of manufacture
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.