Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, 1.0 N Solution (DEA List I Chemical), Spectrum™ Chemical
SDP

Catalog No. 18611058 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-058 20 L Poly Drum
18-611-055 500 mL Amber Glass Bottle
18-611-056 1 L Amber Glass Bottle
18-611-057 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-058 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I12520LTBL

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

Quantity 20 L
Concentration or Composition (by Analyte or Components) 1.0 N ± 0.01 N
Packaging Poly Drum

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.