Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine-125, MP Biomedical

Catalog No. MP016303401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mci
10 mci
2 mci
20 mci
25 mci
5 mci
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP016303401 1 mci
MP016303402 2 mci
MP016303405 5 mci
MP016303410 10 mci
MP016303420 20 mci
MP016303425 25 mci
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP016303401 Supplier MP Biomedicals, Inc Supplier No. 016303401

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
PubChem CID 24841
ChEBI CHEBI:43451
IUPAC Name hydrogen iodide
SMILES I

Specifications

Quantity 1 mci
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.