Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, ACS Grade, ≥ 99.8%, LabChem™

Catalog No. LC155901 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC155901 500 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. LC155901 Supplier LabChem, Inc. Supplier No. LC155901
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Boiling Point 184°C
Melting Point 113°C
Color Purple through Brown
Physical Form Crystals
CAS Max % 100
Quantity 500 g
Identification Passes Test
Packaging Amber Glass
Density 4.93g/cm3
Linear Formula I2
UN Number UN3495
Solubility Information Soluble in water
Formula Weight 253.809
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material Iodine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe dust, vapors.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.