Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Certified, 0.0156N ±0.0001N (0.0078M), LabChem™

Catalog No. LC156022
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156022 1 L Amber Glass
LC156024 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC156022 Supplier LabChem, Inc. Supplier No. LC156022
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Red through Brown
CAS Max % 0.2, 99.39, 0.39, 0.02
Physical Form Liquid
Quantity 1 L
Linear Formula I2
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0156N ±0.0001N (0.0078M)
Identification Passes Test
Packaging Amber Glass
Chemical Name or Material Iodine
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.