Learn More
Specifications
Specifications
| Color | Red through Brown |
| Physical Form | Liquid |
| CAS Max % | 0.32, 99.02, 0.64, 0.02 |
| Quantity | 1 L |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Decomposition Information | Iodine vapor; Potassium oxide |
| Linear Formula | I2 |
| Solubility Information | Soluble in water |
| Formula Weight | 253.809 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.