Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Certified, 12.7%, 1.000N ±0.005N (0.5M), LabChem™

Catalog No. LC156614 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156614 4 L Amber Glass
LC156611 500 mL Amber Glass
LC156612 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. LC156614 Supplier LabChem, Inc. Supplier No. LC156614
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown-Red
Physical Form Liquid
CAS Max % 12.69, 61.91, 25.38, 0.02
Quantity 4 L
Identification Passes Test
Packaging Amber Glass
Linear Formula I2
Solubility Information Soluble in water
Formula Weight 253.809
Percent Purity 12.7%
Grade Certified
Concentration or Composition (by Analyte or Components) 1.000N ±0.005N (0.5M)
Chemical Name or Material Iodine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.