Learn More
Specifications
Specifications
| Color | Brown-Red |
| CAS Max % | 2.54, 92.36, 5.08, 0.02 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Linear Formula | I2 |
| Solubility Information | Soluble in water |
| Formula Weight | 253.809 |
| Percent Purity | 2.5% |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.200N ±0.002N (0.1M) |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Avoid breathing mist, vapors, spray. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If on skin: Wash skin with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Take off contaminated clothing and wash contaminated clothing before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.