Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine concentrate, For 1L standard solution, 0.05 M I2 (0.1 N), Solstice

Catalog No. 60046684 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-684 6 x 1 Ea.
60-032-30 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-046-684 Supplier Solstice Supplier No. 380606X1EA
This item is not returnable. View return policy

for 1L standard solution, 0.05 M I2 (0.1 N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS Min % ≥50.0000%, ≥20.0000%
CAS Max % <70.0000%, <25.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Molecular Formula IK
Linear Formula I2
MDL Number MFCD00011355
UN Number NONH for all modes of transport
Beilstein 3587194
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight 253.81
Packaging Ampoule
Chemical Name or Material Iodine concentrate, For 1L standard solution, 0.05 M I2 (0.1 N)
Show More Show Less
Hazard Statement P305 + P351 + P338-P314
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.