Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Iodine-Iodide), 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. R4000000120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R4000000120 120 mL Amber Glass Bottle
4000-16 500 mL Amber Glass Bottle
4000-32 1 L Amber Glass Bottle
4000-1 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R4000000120 Supplier Ricca Chemical Company Supplier No. 40004
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown
Boiling Point ∼100°C
CAS Min % 1.21, 94.96, 3.82
Melting Point ∼0°C
CAS Max % 1.21, 94.96, 3.82
Density 1.05
Solubility Information Miscible in water
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Iodine (Iodine-Iodide), 0.100 Normal (N/100)
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.