Learn More
Thermo Scientific™ Iodine lump, ultra dry, 99.999% (metals basis)
Specifications
Specifications
| Melting Point | 183-186° subl. |
| CAS | 7553-56-2 |
| Physical Form | Solid |
| Quantity | 25 g |
| Molecular Formula | I{2} |
| MDL Number | MFCD00011355 |
| UN Number | UN3495 |
| Formula Weight | 253.81 |
| Percent Purity | 99.999% |
| Density | 4.930 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.