Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Monochloride Solution, Wijs, for Iodine Absorption Number of Fats and Oils, Ricca Chemical
SDP

Catalog No. R4110000100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R4110000100 100 mL Amber Glass Bottle
411016 500 mL Amber Glass Bottle
411032 1 L Amber Glass Bottle
4110-1 4 L Amber Glass Bottle
13-622-603 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R4110000100 Supplier Ricca Chemical Company Supplier No. 4110100
Add to Cart
Add to Cart

For Analysis of Iodine Absorption Number of Fats and Oils

Chemical Identifiers

CAS 64-19-7 , 7790-99-0 , 7553-56-2
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 96.0, 1.41, 0.09
CAS Max % 100.0, 1.46, 0.1
Grade Laboratory
Packaging Amber Glass Bottle
pH <1
Quantity 100 mL
Chemical Name or Material Iodine Monochloride Solution, Wijs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.