Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Puriss., ≥99.5% (RT), particles (round), Solstice

Catalog No. 60047118 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-118 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-047-118 Supplier Solstice Supplier No. 576551KG

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss.,≥99.5% (RT), particles (round)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Boiling Point 184°C (literature)
Melting Point 113°C (literature)
CAS Max % 100.0000%
Quantity 1 kg
Linear Formula I2
UN Number UN3495
Beilstein 3587194
IUPAC Name diiodine
Formula Weight 253.81
Percent Purity ≥99.5%
Grade puriss.
Packaging Glass Bottle
Density 4.930 g/cm3
Chemical Name or Material Iodine, Puriss., ≥99.5% (RT), particles (round)
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Iodine
EINECSNumber 231-442-4
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.