Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, 99.8+%, ACS reagent

Catalog No. AC423820050
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423820050 5 g Glass Bottle
AC423825000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC423820050 Supplier Thermo Scientific Chemicals Supplier No. 423820050
 Restricted Item
Add to Cart
This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Boiling Point 185°C
Melting Point 113°C
Color Gray
Physical Form Prills
Quantity 5 g
Assay Percent Range 99.8+%
Linear Formula I2
UN Number 3495
Fieser 01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
Merck Index 15, 5057
IUPAC Name diiodine
Formula Weight 253.81
Percent Purity ≥99.80%
Grade ACS Reagent
Packaging Glass Bottle
Vapor Pressure 0.41hPa at 25°C
Density 8.8 (Air = 1.0)
Chemical Name or Material Iodine, 99.8%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh and keep confortable for several minutes.
EINECSNumber 231-442-4
RTECSNumber NN1575000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.