Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, resublimed crystals, Puratronic™, 99.9985% (metals basis)

Catalog No. AA1061936
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1061936 500 g
AA1061988 2.5 g
AA1061914 25 g
AA1061922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AA1061936 Supplier Thermo Scientific Chemicals Supplier No. 1061936
Add to Cart
This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Melting Point 183°C to 186° (sublimation)
Color Black-Blue
Physical Form Solid
Quantity 500 g
Assay Percent Range 99.9985% (Metals basis)
IUPAC Name diiodine
Formula Weight 253.81
Odor Irritating
Appearance Bluish-black crystals
Density 4.93
Chemical Name or Material Iodine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335-H372
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-442-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.