Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Resublimed (DEA List I Chemical), Crystal, USP, 99.8-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18603470 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 25 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-470 25 g Amber Glass Bottle
18-603-472 125 g Amber Glass Bottle
18-603-471 6 x 25 g Amber Glass Bottle
18-603-474 500 g Amber Glass Bottle
18-603-473 6 x 125 g Amber Glass Bottle
18-603-476 2.5 kg Amber Glass Bottle
18-603-475 6 x 500 g Amber Glass Bottle
18-603-477 4 x 2.5 kg Amber Glass Bottle
18-603-478 12 kg Poly Pail
18-603-479 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18-603-470 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I101525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

I1015, 7553-56-2, I2

Spectrum™ Chemical Iodine, Resublimed (DEA List I Chemical), Crystal, USP is a chemical element and it is often found as a contrast material in X-rays, water purification reagent, nutritional supplement, and as an antiseptic . This product is a List I chemical regulated by The United States Drug Enforcement Agency (DEA) which maintains a list of illicit drugs and the chemical materials used to synthetically produce them. List I chemicals are those that are used in the manufacture of the controlled substances and are important to the manufacture of the substances. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 100%
Quantity 25 g
UN Number UN3495
Percent Purity 99.8 to 100.5%
Grade USP
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.