Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Solstice

Catalog No. 6002590 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-90 1 L
60-046-527 6 x 1 L
60-025-91 2.5 L
60-046-526 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-025-90 Supplier Solstice Supplier No. 350901L
Add to Cart
Add to Cart

Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2 , 7681-11-0 , 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 3587194
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
IUPAC Name molecular iodine
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.05 M
Chemical Name or Material Iodine solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.