Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Solstice

Catalog No. 60046527 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-527 6 x 1 L
60-025-90 1 L
60-025-91 2.5 L
60-046-526 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-527 Supplier Solstice Supplier No. 350906X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2 , 7681-11-0 , 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II

Specifications

CAS Min % ≥90.0000%, ≥1.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%, <5.0000%
Quantity 6 x 1 L
Linear Formula I2
Beilstein 3587194
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
IUPAC Name diiodine
Grade Reagent Ph. Eur.
Concentration or Composition (by Analyte or Components) 0.05 M I2 (0.1N)
Packaging Glass Bottle
Chemical Name or Material Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)
Show More Show Less
ShelfLife 360 days from date of manufacture
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.