Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution 0.10N, 0.05M (Potassium Iodide)
SCI_ED

Catalog No. S25937 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 mL
500 mL
Packaging:
0.1N/0.05M
O.1N/0.05M
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25937 10 mL 0.1N/0.05M
S25940 500 mL O.1N/0.05M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25937
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0 , 7553-56-2 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Brown
Physical Form Liquid
Quantity 10 mL
Linear Formula KI
Grade 43% w/v
Concentration or Composition (by Analyte or Components) 0.1N/0.05M
Packaging 0.1N/0.05M
Density 1 to 1.05
Chemical Name or Material Iodine Solution 0.10N, 0.05M (Potassium Iodide)
Safety handling Check Alert storage code Red
Recommended Storage Stable under normal conditions of use and storage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.