Learn More
Description
- Stabilized Iodine is thicker and requires a longer decolorization step or a faster decolorizer
- Stabilized Iodine is less sensitive to light and has a longer shelf life
Specifications
Specifications
| Includes | 3.7 L |
| Format | Liquid |
| Boiling Point | 184°C |
| Storage Requirements | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
