Learn More
Promega Iodoacetamide, 15mg
Catalog No. PRVB1010 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
15 mg
Catalog No. Quantity
PRVB1010 15 mg
1 options
Catalog No. PRVB1010 Supplier Promega Supplier No. VB1010
AccuMAP Low pH Protein Digestion Kit enables suppression of sample preparation-induced artificial deamidation, disulfide bond scrambling and oxidation overthe course of sample preparation. Pre-existing non-enzymatic modifications remain intact.
  • Suppress Artificial Deamidation and Disulfide Bond Scrambling in Protein Sample Prep
  • Sample preparation completed in 4.5−5 hours
  • Highly reproducible digestion results
  • Use for reduced and nonreduced proteins
Specifications
Content And Storage <-65°C
Quantity 15 mg