Description
- Ultrapure, proteomic-grade iodoacetamide is used for efficient alkylation of thiols while minimizing oxidation of the competing thiol pairs in protein sample
Specifications
Specifications
| For Use With (Application) | For efficient alkylation of thiols while minimizing oxidation of competing thiol pairs in Protein samples |
| Quantity | 25000 mg |