Learn More
Abcam Iodoacetamide, Alkylating Agent, Solid, >98% Purity, 25 g
Description
- Molecular weight: 184.96 g/mol
- Purity: >98%
- Form: Solid
- Composition: 100% Iodoacetamide
- Properties: Alkylating agent used in peptide mapping; covalently binds to cysteine thiols; irreversible inhibitor of enzymes with cysteine at the active site; reacts with histidine residues.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Target Molecule | Thiols (Cysteine), Histidine |
| Quantity | 25 g |
| For Use With (Application) | Peptide mapping |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.