Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoacetic Anhydride, 97%

Catalog No. AC307590010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC307590010 1 g
AC307592500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC307590010 Supplier Thermo Scientific Chemicals Supplier No. 307590010
  • $333.00
    $266.65 / Each of 1

    Save $66.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 54907-61-8 | C4H4I2O3 | 353.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54907-61-8
Molecular Formula C4H4I2O3
Molecular Weight (g/mol) 353.88
MDL Number MFCD00001080
InChI Key RBNSZWOCWHGHMR-UHFFFAOYSA-N
Synonym iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
PubChem CID 2733254
IUPAC Name (2-iodoacetyl) 2-iodoacetate
SMILES C(C(=O)OC(=O)CI)I

Specifications

Melting Point 43.0°C to 49.0°C
Color Yellow
Flash Point >110°C
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula (ICH2CO)2O
Quantity 1 g
UN Number 3261
Formula Weight 353.88
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Iodoacetic Anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.