Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene 99.0+%, TCI America™
SDP

Catalog No. I0050100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0050100G 100 g
I005025G 25 g
I0050500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0050100G Supplier TCI America Supplier No. I0050100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 188°C
Quantity 100 g
Formula Weight 204.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodobenzene
EINECSNumber (9)-1501
RTECSNumber DA3390000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.