Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodocyclohexane (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. I0056100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0056100G 100 g
I005610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0056100G Supplier TCI America Supplier No. I0056100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 626-62-0
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD00003826
InChI Key FUCOMWZKWIEKRK-UHFFFAOYSA-N
Synonym cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
PubChem CID 12289
IUPAC Name iodocyclohexane
SMILES IC1CCCCC1

Specifications

Boiling Point 180°C
Quantity 100 g
Formula Weight 210.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodocyclohexane (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.