Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoform, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18603494 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-494 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-603-494 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I203125KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

I2031, 75-47-8, CHI3

Spectrum™ Chemical Iodoform, USP is a light yellow substance with a pungent odor and similar to chloroform and is an organoiodine compound. Iodoform has been used as a disinfectant. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Iodoform, USP, 99-100.5%, I2031, CHI3

Specifications

CAS 75-47-8
CAS Max % 100%
Boiling Point 218°C/424.4°F
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Poly Pail
Molecular Formula CHI3
Quantity 25 kg
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILES IC(I)I
IUPAC Name triiodomethane
Molecular Weight (g/mol) 393.73
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Iodoform
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.