Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodopentafluorobenzene, 99%, stabilised over copper

Catalog No. AC312200050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312200050 5 g Glass bottle
AC312200010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312200050 Supplier Thermo Scientific Chemicals Supplier No. 312200050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 827-15-6 | C6F5I | 293.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-15-6 , 7440-50-8
Molecular Formula C6F5I
Molecular Weight (g/mol) 293.96
InChI Key OPYHNLNYCRZOGY-UHFFFAOYSA-N
Synonym iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
PubChem CID 70008
IUPAC Name 1,2,3,4,5-pentafluoro-6-iodobenzene
SMILES C1(=C(C(=C(C(=C1F)F)I)F)F)F

Specifications

Color Colorless
Density 2.2040g/mL
Boiling Point 162.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.496
Quantity 5 g
Specific Gravity 2.204
Formula Weight 293.96
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Iodopentafluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 212-565-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.