Learn More
Ion 520™ Chip Kit



Description
The Ion 520 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL Sequencing Systems. The Ion 520 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times in as little as 2.5 hours for 200 bp sequencing.
The Ion 520 Chip Kit offers:
• Sample tracking functionality from an integrated barcode
• A rapid run time of only 2.5–4 hours, generating 3–6 million reads, depending on application
• Read lengths up to 600 bp to support a variety of research applications, including targeted gene sequencing and microbial sequencing
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 510 & Ion 520 & Ion 530 Kit-Chef, the Ion 520 & Ion 530 ExT Kit-Chef, and the Ion 520 & Ion 530 Kit-OT2.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 8 chips |
| Product Line | Ion 520 |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion S5™ XL System |
| Number of Reads per Chip | 3-6M |
| Product Type | Chip Kit |
| Shipping Condition | Room Temperature |
| Content And Storage | Store at room temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.