Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion 530™ Chip Kit
GSA_VAGREENER_CHOICE

Catalog No. A27764
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 chips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27764 8 chips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27764 Supplier LIFE TECHNOLOGIES Supplier No. A27764
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Ion 530 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL Sequencing Systems. The Ion 530 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of less than 2.5 hours for 200 bp sequencing.

The Ion 530 Chip offers:

• Sample tracking functionality from an integrated barcode
• A rapid run time of only 2.5–4 hours, generating 15–20 million reads
• Read lengths up to 600 bp to support a variety of research applications, including targeted gene sequencing and microbial sequencing
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods

Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 510 & Ion 520 & Ion 530 Kit-Chef, the Ion 520 & Ion 530 ExT Kit-Chef, and the Ion 520 & Ion 530 Kit-OT2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Ion 530
Content And Storage Store at room temperature
For Use With (Equipment) Ion GeneStudio S5 System, Ion S5™ XL System
Number of Reads per Chip 15-20M
Product Type Chip Kit
Quantity 8 chips
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.