Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion 540™ Chip Kit
GREENER_CHOICE

Catalog No. A27765
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 Chips
8 Chips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27765 4 Chips
A27766 8 Chips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27765 Supplier LIFE TECHNOLOGIES Supplier No. A27765
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Ion 540™ Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 540 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 2.5 hours for 200 bp sequencing.

The Ion 540 Chip Kit offers:

• Sample tracking functionality from an integrated barcode
• A rapid run time of as little as 2.5 hours for 200 bp sequencing, generating 60–80 million reads
• Support for a variety of research applications, including sequencing of up to three human exomes, transcriptome profiling, and targeted resequencing of gene panels
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods

Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 540 Kit-Chef and the Ion 540 Kit-OT2.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Ion 540
Content And Storage Store at room temperature
Format Kit
For Use With (Application) Sequencing
Number of Reads per Chip 60-80M
Product Type Chip Kit
Quantity 4 Chips
Sequencing Type Genome and DNA Sequencing
Shipping Condition Room Temperature
Species Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.