Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion 540™ Kit-Chef (2 sequencing runs per initialization)

Catalog No. A30011
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30011 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30011 Supplier LIFE TECHNOLOGIES Supplier No. A30011
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Ion 540™ Kit-Chef enables robust and automated template preparation and sequencing of up to 200 base-read libraries using the Ion Chef™ and the Ion S5™ or Ion S5™ XL Sequencing Systems. The Ion 540 Kit contains pre-packaged template and sequencing reagent cartridges with integrated sample tracking, delivering an automated workflow for sequencing of 8 loaded chips with complete run traceability. The kit contains sufficient reagents and supplies for 4 initializations and 8 sequencing runs on the Ion S5 or Ion S5 XL systems.

The Ion 540 Kit for the Ion Chef and Ion S5/Ion S5 XL workflow:
• Provides template preparation and chip loading reagents for up to 200 base-read libraries
• Simplifies your research with ready-to-use, disposable reagent cartridges for sequencing
• Delivers fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
• Leverages the latest advancements in data quality and accuracy

The Ion S5/Ion S5 XL sequencers, along with the Ion OneTouch™ 2 System, provide reproducible sequencing workflows. Delivering up to 80 million reads in a single run, the Ion 540 Chip empowers rapid exome and transcriptome analysis. Combined with Ion AmpliSeq™ technology and Ion Reporter™ Software for data interpretation, the Ion S5/Ion S5 XL systems provide a cost-effective and comprehensive solution for your translational or inherited disease research needs.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Ion 540
Content And Storage • Ion S5 Chef Solutions, store at room temperature
• Ion S5 Chef Supplies, store at room temperature
• Ion 540 Chef Reagents, store at -5°C to -30°C
• Ion S5 Sequencing Solutions, store at room temperature
• Ion S5 Sequencing Reagents, store at -5°C to -30°C
Product Type Chef Kit
Quantity 8 reactions
Shipping Condition Qualified for Ambient or Wet Ice
Workflow Step Template Preparation
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.