Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion AmpliSeq™ Exome RDY Kit 1x8

Catalog No. A38262
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A38262 8 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A38262 Supplier Ion Torrent™ Supplier No. A38262
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Line Ion AmpliSeq
Software Type Torrent Suite Software, Ion Reporter Software
Starting Material DNA
Content And Storage • Ion AmpliSeq Exome RDY Panel 1x8
• Ion AmpliSeq Library Kit Plus

Format 96-well Plate
For Use With (Equipment) Ion GeneStudio S5 System
Libraries Targeted Sequencing Library
Product Type Exome RDY Kit
Quantity 8 Reactions
Sequencing Type Genome & DNA Sequencing
Shipping Condition Approved for shipment on Wet or Dry Ice
Workflow Step Library Generation
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.