Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion AmpliSeq™ Library Kit 2.0
GSA_VA

Catalog No. 4480441
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
384 Reactions
96 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
44-804-41 96 Reactions
44-804-42 384 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-804-41 Supplier Ion Torrent™ Supplier No. 4480441
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.

The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.

Scalable Multiplex PCR Reactions
The Ion AmpliSeq 2.0 technology enables scalable multiplex PCR reactions from 12- to 24,000-plex in a single well using just 10 ng of starting DNA. Ion AmpliSeq panels and primer pools allow highly multiplexed PCR amplification of thousands of genomic target regions, with superior coverage uniformity and specificity, without the need of a specialized microfluidics PCR platform. In addition, the primers contain proprietary modifications that enable removal of primer sequences during library preparation, for efficient target assessment during sequencing. Multiple primer pools can be used to create overlapping amplicons that enable complete coverage of large targets. Ion AmpliSeq™ Custom Primer Pools are designed via the Ion AmpliSeq™ Designer Tool, available at www.ampliseq.com.

Barcoded Library Preparation
The Ion AmpliSeq Library Kit 2.0 includes reagents for generating amplicons with Ion AmpliSeq primers and preparing libraries from the resulting amplicons. The kit enables the preparation of barcoded libraries using Ion Xpress™ Barcode Adapters 1-96 kits or IonCode™ Barcode Adapters 1-384 kits. Barcoded libraries can be combined and loaded onto a single Ion chip to minimize the sequencing run time and cost and allow for accurate sample-to-sample comparisons.

Easy and Flexible Protocol
The Ion AmpliSeq Library Kit 2.0 uses a plate-based format for easier sample handling and tracking, and for compatibility with automation and high-throughput laboratories. The library preparation uses very small quantities of starting material per PCR. The resulting DNA libraries are ready for downstream template preparation using the automated Ion Chef™ System (which also provides the option of automated library preparation using the Ion AmpliSeq™ Kit for Chef DL8 (Cat. No. A29024)) or the Ion OneTouch™ 2 System followed by sequencing. The intuitive Torrent Suite™ Software and optional Ion Reporter™ Software enables automation of data analysis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 7 tubes, store at -10°C to -30°C
• 1 tube Low TE Buffer, store at room temperature
Technique Library Generation
For Use With (Equipment) Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
High-throughput Compatibility High-throughput Compatible, Multiplexing
Libraries Targeted Sequencing Library
Multiplex Capability 12- to 3072-plex per well
Product Line Ion AmpliSeq
Product Type Library Kit 2.0
Quantity 96 reactions
Sequencing Type Genome & DNA Sequencing
Species Human
For Use With (Application) Next-Generation Sequencing (NGS)
No. of Reactions 96 Reactions
Sample Type DNA (Genomic), DNA from FFPE Tissues, Tissue (FFPE)
Starting Material 10 ng gDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.