Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion Library Equalizer™ Kit

Catalog No. 4482298
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-822-98 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-822-98 Supplier Ion Torrent™ Supplier No. 4482298
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Sequencing Type Epigenetic Sequencing, Genome & DNA Sequencing
For Use With (Application) Amplicon Sequencing
Starting Material DNA
Quantity 96 reactions
Product Line Equalizer
Format 96-well Plate
For Use With (Equipment) Ion PGM™ System, Ion Proton™ System, Ion Personal Genome Machine (PGM™) System
Libraries Fragment Library, Targeted Sequencing Library, Normalized Library
Multiplex Capability No Limit on Multiplexing
Product Type Library Equalizer Kit
Sample Type Any Sample Type
Sample Volume Fragment or Targeted AmpliSeq Libraries
Species All Species
Throughput Up to 10 Samples
Workflow Step Library Barcoding, Library Generation, Template Preparation
Content And Storage • Equalizer Primers - store at 2–8°C
• Equalizer Capture - store at 2–8°C
• Equalizer Wash Buffer - store at room temperature (can be stored at 2–8°C)
• Equalizer Elution Buffer - store at 2–8°C
• Equalizer Beads - store at 2–8°C

Note kits are shipped at ambient temperature store each component at specified temperatures.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.