Learn More
Abcam ION NaTRIUM Green™-2 TMA+ Salt, Cytosolic Na+ Indicator, 500 μg
Description
- Molecular Weight: 1087.19 g/mol
- Purity: >90%
- Description: Cell membrane impermeable cytosolic Na+ indicator
- Solubility: Water soluble
- Fluorescence: Excitation maximum at 517 nm; fluorescent probe
- Activity: Active in vitro
- Form: Solid
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Sodium |
| Shipping Condition | Ambient |
| Label or Dye | Fluorescent Probe |
| Form | Solid |
| Detection Method | Fluorescence |
| Detection Location | In-Solution Detection |
| Content And Storage | Store at -20°C. |
| Buffer | 90% ION NaTRIUM Green™-2 TMA+ salt |
| Formulation | 90% ION NaTRIUM Green™-2 TMA+ Salt |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.