Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ION Potassium Green-2 AM, K+ Indicator, Solid, 500 ug

Catalog No. 502583311
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3311 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3311 Supplier Abcam Supplier No. AB142806500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell membrane permeable fluorescent K+ indicator.

  • Molecular weight: 1128.02 g/mol
  • Purity: >90%
  • Form: Solid
  • Properties: Cell membrane permeable K+ indicator, fluorescent probe
  • Mechanism: Works in cells with non-specific esterases to hydrolyse AM to release dye
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Potassium
Substrate Potassium
Shipping Condition Ambient
Kit Contents 500 μg ION Potassium Green-2 AM
Label or Dye Fluorescent Probe
Detection Location In-Cell Detection
Form Solid
Detection Method Fluorescence
Alternate Part Number ab142807
Buffer 90% ION Potassium Green-2 AM
Content And Storage Store at -20°C.
Formulation 90% ION Potassium Green-2 AM
Sample Type Cells
Quantity 500 μg
For Use With (Application) Potassium detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.