Learn More
Ion Torrent™ Ion Reporter™ Server System

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Software Type | Informatics Tools |
| Quantity | 1 each |
| Product Line | Ion Reporter Software |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion PGM™ System, Ion Proton™ System, Ion S5™ XL System |
| Operating System | Ubuntu |
| Software Category | Analysis |
| Content And Storage | • Ion Reporter Software, includes first year software license • 1 Dell T440 computer server • Field installation of computer server and software |
| Description | Ion Reporter Server • Latest release version: 5.20 • Server shipping version: 5.16 |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.