Learn More
Ion Torrent™ Ion S5™ 2X Cleaning Solution
Description
Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.
Features include:
- The ability to run a pulse clean in the event of a solid-state buffer leak
- The option to deep clean the fluidics of the Ion S5 Sequencer
Specifications
Specifications
| Quantity | 2 Bottles |
| For Use With (Equipment) | Ion GeneStudio S5 and S5 |
| Height (Metric) | 131.1 mm |
| Length (Metric) | 61 mm |
| Product Type | 2X Cleaning Solution |
| Width (Metric) | 61 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.