Learn More
Ion S5™ Precision ID Chef & Sequencing Kit (1 run per initialization)
Description
The Ion S5 Precision ID Chef & Sequencing Kit enables robust and automated template preparation and sequencing of Precision ID libraries using the Ion GeneStudio S5 worfklow (eight initializations and eight sequencing runs). This kit provides template preparation and chip loading reagents for libraries generated with commercially available Precision ID panels (GlobalFiler NGS STR Panel v2, Identity Panel, Ancestry Panel, mtDNA Whole Genome Panel, mtDNA Control Region Panel, or custom Ion AmpliSeq SNP panel). For four initializations and eight sequencing runs, see Cat. No. A33208.
The Ion S5 Precision ID Chef & Sequencing Kit for the Ion Chef and Ion GeneStudio S5 workflow:
- Provides template preparation and chip loading reagents for Precision ID libraries
- Simplifies your testing with ready-to-use, disposable reagent cartridges for sequencing
- Delivers fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
- Leverages our latest advancements in data quality and accuracy
- Maximizes flexibility through compatibility with both the Ion 520 and Ion 530 chips
The Ion GeneStudio S5 sequencers, along with the Ion Chef System, provide reproducible sequencing workflows with minimal hands-on time. Choose to pair the Ion 520 & Ion 530 Kit with the Ion 520 (up to 5M reads) or Ion 530 (up to 20M reads) Chip kit, allowing for flexibility of total throughput.
Specifications
Specifications
| Quantity | 8 Reactions |
| Product Line | Ion S5 |
| For Use With (Equipment) | Ion Chef, Ion GeneStudio S5 |
| Product Type | Precision ID Chef and Sequencing Kit |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Content And Storage | • Ion S5 Precision ID Chef solutions, store at room temperature • Ion S5 Precision ID Chef supplies, store at room temperature • Ion S5 Precision ID Chef reagents, store at -5°C to -30°C • Ion S5 Precision ID sequencing solutions, store at room temperature • Ion S5 Precision ID sequencing reagents, store at -5°C to -30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.