Learn More
Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization)
Description
The Ion S5 Precision ID Chef & Sequencing Kit enables robust and automated template preparation and sequencing of Precision ID libraries using the Ion Chef System and an Ion GeneStudio S5 sequencing system (four initializations and eight sequencing runs). Using this kit with the Ion S5 workflow provides template preparation and chip loading reagents for libraries generated with commercially available Precision ID panels (GlobalFiler NGS STR Panel v2, Identity Panel, Ancestry Panel, mtDNA Whole Genome Panel, mtDNA Control Region Panel, or custom Ion AmpliSeq SNP panel). For eight initializations and eight sequencing runs, see Cat. No. A35850.
The Ion S5 Precision ID Chef & Sequencing Kit for the Ion Chef and Ion GeneStudio S5 workflow:
- Provides template preparation and chip loading reagents for Precision ID libraries
- Simplifies your testing with ready-to-use, disposable reagent cartridges for sequencing
- Delivers fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
- Leverages our latest advancements in data quality and accuracy
- Maximizes flexibility through compatibility with both the Ion 520 and Ion 530 chips
The Ion GeneStudio S5 sequencers, along with the Ion Chef System, provide reproducible sequencing workflows with minimal hands-on time. A single Ion 530 Chip accommodates 32 libraries made with the GlobalFIler NGS STR Panel v2.
Specifications
Specifications
| Quantity | 8 reactions |
| Product Line | Ion S5 |
| Product Type | Precision ID Chef and Sequencing Kit |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Content And Storage | • Ion S5 Precision ID Chef solutions, store at room temperature • Ion S5 Precision ID Chef supplies, store at room temperature • Ion S5 Precision ID Chef reagents, store at -5°C to -30°C • Ion S5 Precision ID sequencing solutions, store at room temperature • Ion S5 Precision ID sequencing reagents, store at -5°C to -30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.