Learn More
World Precision Instrument Ion Selective Electrode - Potassium
SDP
Supplier:  World Precision Instrument KWIKPOT2
Encompass_Preferred
Accurately measure potassium
These highly stable electrodes ac­cu­rate­ly measure cal­ci­um, po­tas­si­um, hydrogen and TPP (Tetraphenylphosphonium) ion activity. Tips consist of 2 mm di­am­e­ter plas­tic tubes sealed at one end with an ion-sen­si­tive mem­brane. After filling with elec­tro­lyte so­lu­tion, you can in­sert the tube into the hold­er and con­nect it to a pH meter. Tips and hold­ers are in­ter­change­able, so one tip may be re­placed with an­oth­er sensitive to a dif­fer­ent ion. Replac­ing a tip takes less than a minute. Elec­trode tips nor­mal­ly last sev­er­al months, when stored proper­ly in sa­line so­lu­tion. When re­place­ment is neces­sary, only the tip needs be re­placed.
Catalog No. 50-252-1153
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.